Order Entry
Austria
ContactUsLinkComponent
Bovine native Ceramide II (from Brain)
Bovine native Ceramide II (from Brain)
Catalog # ENZOALX303002M025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Bovine native Ceramide II (from Brain)
Catalog # ENZOALX303002M025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Protein/peptide type:
    Native
  • Source:
    Brain
  • Species:
    Bovine
  • Seq:
    02
  • Protein/peptide name:
    Ceramide II
  • Purity:
    >98%

Specifications

About this item

Prepared from cerebrosides by a modified protocol of H.E. Carter et al.; J. Lipid Res. 2, 228 (1961).

  • Contains α-hydroxy fatty acids

Semisynthetic (from bovine brain cerebrosides).