Bovine native Ganglioside GQ1b . tetrasodium salt (from Brain)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Specifically promotes neural differentiation of human neuroblastoma cells.
Specifications
- Protein/peptide type:Native
- Protein/peptide name:Ganglioside GQ1b . tetrasodium salt
- Purity:>98% (TLC)
- Source:Brain
- Species:Bovine
Specifications
Frequently Bought Together