Order Entry
Austria
ContactUsLinkComponent
Bovine native Ganglioside GD3 . disodium salt (from Brain)
Bovine native Ganglioside GD3 . disodium salt (from Brain)
Supplier:  Enzo Life Sciences
Certificates
undefined
Bovine native Ganglioside GD3 . disodium salt (from Brain)
Supplier:  Enzo Life Sciences

Specifications

  • Protein/peptide type:
    Native
  • Source:
    Brain
  • Species:
    Bovine
  • Protein/peptide name:
    Ganglioside GD3 . disodium salt
  • Purity:
    >98% (TLC)

Specifications

Frequently Bought Together