Bovine native Ganglioside GD1b . disodium salt (from brain)
Catalog # ENZOALX302009M001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Native
- Source:Brain
- Species:Bovine
- Seq:02
- Protein/peptide name:Ganglioside GD1b . disodium salt
- Purity:>98% (TLC)