Ac-IETD-AFC
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Caspase and granzyme substrate
Fluorogenic substrate for caspase-8/caspase-3 processing enzyme and granzyme B. Also a substrate for caspase-10. The IETD sequence is based on caspase-3 proenzyme cleavage site.
Specifications
- Protein/peptide name:Ac-IETD-AFC
- Purity:>98% (HPLC)