Order Entry
Austria
ContactUsLinkComponent
Ac-LEVD-AMC
Ac-LEVD-AMC
Catalog # ENZOALX260083M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Ac-LEVD-AMC
Catalog # ENZOALX260083M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    04
  • Protein/peptide name:
    Ac-LEVD-AMC
  • Purity:
    >99% (HPLC)

Specifications

About this item

Caspase-4 substrate

Fluorogenic substrate for caspase-4.