Ac-DMQD-AMC
Catalog # ENZOALX260078M005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:03
- Protein/peptide name:Ac-DMQD-AMC
- Purity:>98% (HPLC)
Specifications
About this item
Caspase-3 substrate
Fluorogenic substrate for caspase-3.