Ac-VEID-AMC
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Caspase-6 substrate
Fluorogenic substrate for caspase-6 (Km=30µML, Kcat/Km=168000M-1sec-1) and related cysteine proteases. Sequence is based on lamin A site of cleavage. Ex.:340 to 360 nm, Em.:440 to 460 nm.
Specifications
- Protein/peptide name:Ac-VEID-AMC
- Purity:>97% (HPLC)