Order Entry
Austria
ContactUsLinkComponent
Ac-DEVD-AMC
Ac-DEVD-AMC
Supplier:  ENZO LIFE SCIENCES
Certificates
Ac-DEVD-AMC
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide name:
    Ac-DEVD-AMC
  • Purity:
    >97% (HPLC)
  • Formulation:
    Lyophilized

Specifications

Frequently Bought Together