Human recombinant Control:Fc fusion protein
Catalog # ENZOALX203004C050
Lieferant: Enzo Life Sciences
CAS-Nummer:
Spezifikationen
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Reconstitution instructions:Reconstitute with 50 µl sterile distilled water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
- Protein/peptide name:Control:Fc fusion protein
- Purity:>90% (SDS-PAGE)
- Concentration:1 mg/ml (after reconstitution).
- Formulation:Lyophilised. Contains PBS.
Spezifikationen
Über diesen Artikel
Negative control for Fc chimeric proteins
Produced in HEK 293 cells. The extracellular domain of mutated hThy-1, hThy-1 (RLE) (aa 20-130), is fused to the Fc portion of human IgG1. The integrin binding sequence RLD of hThy-1 has been changed to RLE abolishing the binding of integrins.
Negative control for Fc chimeric proteins. Does not bind to a series of ligands in an ELISA assay. Does not inhibit binding between the thymoma cell line EL-4 expressing β3-integrin and astrocytes expressing high level of Thy-1 in mice.