Human recombinant DESC1
Catalog # ENZOALX2017511
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 Vial
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Biological activity:Trypsin-like serine protease.
- Protein/peptide name:DESC1
- Concentration:100 nM
- Formulation:Liquid. In 100 mM TRIS, pH 9,0, containing 200 mM sodium chloride and 10% glycerol.
Specifications
About this item
Produced in Drosophila Schneider 2 (S2) cells. Recombinant human DESC1 (aa 44-423) is fused at the C-terminus to a V5-His-tag.