Order Entry
Austria
ContactUsLinkComponent
Human recombinant alpha-Synuclein (from E. coli)
Human recombinant alpha-Synuclein (from E. coli)
Catalog # ENZOALX201265C100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant alpha-Synuclein (from E. coli)
Catalog # ENZOALX201265C100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    Alpha-Synuclein
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    1 mg/ml
  • Formulation:
    Liquid. In 20 mM TRIS-HCl, pH 7,5, containing 0,1M sodium chloride and 1 mM magnesium dichloride.

Specifications

About this item

Produced in E. coli. alpha-synuclein is a major component of Parkinson's disease aggregates and is implicated in the pathogenesis of Parkinson's disease and related neurodegenerative disorders.

alpha-synuclein accumulates in the brains of sporadic Parkinson's disease patients as a major component of Lewy bodies, which are intraneuronal cytoplasmic inclusions characteristic of Parkinson's disease.

alpha-synuclein appears to associate with other proteins that aggregate and is found in beta-amyloid plaques and neuritic tangles in Alzheinmer's disease.