Human recombinant PARP1 (from Insect cells (Sf9)), HIS Tag
Catalog # ENZOALX201250C010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:PARP1
- Purity:>98% (SDS-PAGE)
- Concentration:0,5 mg/ml
- Formulation:Liquid. In 50 mM TRIS-HCl, pH 7,5, containing 100 mM sodium chloride, 50 mM imidazole, 0,2% NP-40 and 10% glycerol.
- Grade:High purity
Specifications
About this item
Produced in Sf21 cells. Human full-length PARP-1 is fused to a Myc-tag and a His-tag.