Order Entry
Austria
ContactUsLinkComponent
Recombinant dank (from E. coli)
Recombinant dank (from E. coli)
Catalog # ENZOALX201143C025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Recombinant dank (from E. coli)
Catalog # ENZOALX201143C025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Seq:
    02
  • Protein/peptide name:
    Dank
  • Purity:
    >99%
  • Concentration:
    2,8 mg/ml
  • Formulation:
    Liquid. In 20 mM TRIS/HCl, pH 7,2, containing 100 mM NaCl, 40 mM KCl, 0,1 mM EDTA, 1 mM DTT and 5% glycerol.

Specifications

About this item

Produced in E. coli.