Mouse recombinant CD14 (from CHO cells), HIS Tag
Catalog # ENZOALX201134C010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Mouse
- Seq:02
- Tag sequence:HIS Tag
- Biological activity:Inhibits binding of LPS to CD14. Up to 5 µg/ml mouse CD14 inhibit binding of FITC-LPS (0,5 µg/ml) to mouse CD14+ CHO transfectants (FACS).
- Reconstitution instructions:Reconstitute with 40 µl sterile, ultra - pure distilled water. This solution can then be diluted with PBS or other buffers.
- Protein/peptide name:CD14
- Purity:90-95% (SDS-PAGE)
- Concentration:1 mg/ml after reconstitution.
- Formulation:Lyophilised from PBS, pH 7,2.
- Purification:His-tag-affinity purified
Specifications
About this item
Produced in CHO cells.