Human recombinant KillerTRAIL™ (soluble) (from E. coli)
Produced in E. coli. The extracellular domain of human TRAIL (aa 95-281) is fused at the N-terminus to a His-tag and a linker peptide.
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Protein/peptide name:KillerTRAIL™ (soluble)
- Biological activity:Induces apoptosis in a concentration range of 10 - 100 ng/ml.Note: Does not require a cross-linking enhancer for its potent biological activity. For cell lines that require extensive cross-linking of the TRAIL-Rs for killing (e.g. Jurkat) use SuperKillerTRAI.
- Reconstitution instructions:For a special storage and dilution buffer see KillerTRAIL™ Storage and dilution buffer (Prod. No. ALX - 505 - 005).
- Purity:>99% (SDS-PAGE)
- Formulation:Liquid. In 20 mM HEPES, pH 7,4, 300 mM sodium chloride, 0,01% Tween 20, 1% sucrose and 1 mM DTT.
- Purification:Affinity purified. Sterile filtered
Frequently Bought Together