Human recombinant PARP1 (from Insect cells (Sf9))
Catalog # ENZOALX201063C020
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Seq:02
- Protein/peptide name:PARP1
- Purity:>99% (SDS-PAGE)
- Concentration:1 mg/ml
- Formulation:Liquid. In 100 mM TRIS-HCl, pH 7,5, containing 14 mM β-mercaptoethanol, 0,5 mM EDTA, 0,5 mM PMSF and 10% glycerol.
- Grade:High purity
- Purification:Affinity purified
Specifications
About this item
Produced in Sf9 cells.