Order Entry
Austria
ContactUsLinkComponent
Human recombinant MMP3 (catalytic domain) (from E. coli)
Human recombinant MMP3 (catalytic domain) (from E. coli)
Catalog # ENZOALX201042C005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant MMP3 (catalytic domain) (from E. coli)
Catalog # ENZOALX201042C005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    MMP3 (catalytic domain)
  • Concentration:
    0,1 mg/ml
  • Formulation:
    Liquid. In 50 mM Tris-HCl, pH 7,5, containing 10 mM CaCl₂, 1 µM ZnCl₂ and 0,05% sodium azide.
  • Purification:
    No other MMP contaminants are detectable

Specifications

About this item

Produced in E. coli.

Highly purified recombinant enzyme containing the 22 kDa catalytic domain of MMP-3.