Order Entry
Austria
ContactUsLinkComponent
L-TAT control peptide, FITC (Fluorescein Isothiocyanate)
L-TAT control peptide, FITC (Fluorescein Isothiocyanate)
Catalog # ENZOALX168009FR050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
L-TAT control peptide, FITC (Fluorescein Isothiocyanate)
Catalog # ENZOALX168009FR050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 µl
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Seq:
    02
  • Protein/peptide name:
    L-TAT control Peptide
  • Purity:
    >90% (HPLC)
  • Concentration:
    1 mM
  • Formulation:
    Liquid. In sterile PBS.
  • Tested applications:
    Control Peptide

Specifications

About this item

Control peptide for L-JNKI

Control peptide for L-JNKI. For more details see L-JNKI1 (Prod.No. BML-EI354).