Order Entry
Austria
ContactUsLinkComponent
Bovine;dog;human;mouse;pig;rat endothelin 1
Bovine;dog;human;mouse;pig;rat endothelin 1
Supplier:  ENZO LIFE SCIENCES
Certificates
Bovine;dog;human;mouse;pig;rat endothelin 1
Supplier:  ENZO LIFE SCIENCES

Specifications

  • CAS No.:
    117399-94-7
  • Species:
    Pig
    Human
    Bovine
    Dog
    Rat
    Mouse
  • Protein synonyms:
    ET-1
  • Protein/peptide name:
    Endothelin 1
  • Purity:
    >98% (HPLC)
  • Solubility:
    1 mg/ml soluble in DMSO or 5% acetic acid
  • Sequence:
    H-Cys-Ser-Cys-Ser-Ser-Leu-Met-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-His-Leu-Asp-Ile-Ile-Trp-OH
    (Disulfide bonds between Cys1-Cys15and Cys3-Cys11)
  • Formulation:
    Lyophilised
  • Molecular formula:
    C₁₀₉H₁₅₉N₂₅O₃₂S₅

Specifications

Frequently Bought Together