Blocking peptide for Adiponectin receptor 1 pAb Blocking Peptide
Catalog # ENZOALX151045C100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 µG
- Seq:02
- Protein/peptide name:Blocking Peptide for Adiponectin receptor 1 pAb Blocking Peptide
- Purity:>90% (HPLC)
- Concentration:3,5 mg/ml
- Formulation:Liquid. In 20 mM acetate buffer, pH 4,5.
Specifications
About this item
Synthetic peptide corresponding to aa 345-369 of human, mouse or rat adiponectin receptor 1 (AdipoR1).