Order Entry
Austria
ContactUsLinkComponent
Angiotensin I/II (1-7)
Angiotensin I/II (1-7)
Catalog # ENZOALX151041M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Angiotensin I/II (1-7)
Catalog # ENZOALX151041M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    02
  • Protein/peptide name:
    Angiotensin I/II (1-7)
  • Purity:
    >98% (HPLC)
  • Formulation:
    TFA salt

Specifications

About this item