Order Entry
Austria
ContactUsLinkComponent
Human recombinant HSP40/Hdj1 (from E. coli)
Human recombinant HSP40/Hdj1 (from E. coli)
Supplier:  ENZO LIFE SCIENCES
Certificates
Human recombinant HSP40/Hdj1 (from E. coli)
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein/peptide name:
    HSP40/Hdj1
  • Purity:
    >80% (SDS-PAGE)
  • Formulation:
    Liquid, in Dulbecco’s PBS
  • Purification:
    Purified by multi-step chromatography

Specifications

Frequently Bought Together