Bovine native gamma-Crystallin (from Eye lens)
Catalog # ENZOADISPP240J
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Native
- Source:Eye lens
- Species:Bovine
- Seq:02
- Protein/peptide name:gamma-Crystallin
- Purity:>90% (SDS-PAGE; Western blot)
- Formulation:Liquid, in PBS containing 0,09% sodium azide
- Purification:Purified by multi-step chromatography
Specifications
About this item
Isolated from bovine eye lens.