Human Recombinant EGF (from E. coli)
Catalog # ENZOADI908060100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:EGF
- Purity:>98% (SDS-PAGE; RP-HPLC)
- Formulation:Lyophilized. Sterile filtered liquid from a concentrated solution with no additives. It is recommend to reconstitute this protein in sterile 18 MΩ-cm H₂O not less than 100 µg/ml.
- Purification:Purified by multi-step chromatography.
Specifications
About this item
Produced in E.coli.