FluoForte™ Reagent
Catalog # ENZOENZ52015
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 mg
- Description:FluoForte™ reagent
- Seq:02
- Purity:≥95% (HPLC)
- Size:1 mg
Specifications
About this item
Calcium dye
Used in a homogeneous fluorescence-based assay for detecting intracellular calcium mobilization across a broad spectrum of biological targets. Fluorescent calcium indicator dye developed to improve cell loading and calcium response while maintaining the convenient Fluo-3, Fluo-4 and Calcium 4 spectral wavelength of maximum excitation at ~490 nm and maximum emission at ~520 nm. Relative to the cited dyes, FluoForte™ AM dye yields the brightest signal and largest assay window.