Order Entry
Austria
ContactUsLinkComponent
AMC (7-Amino-4-methylcoumarin) standard
AMC (7-Amino-4-methylcoumarin) standard
Catalog # ENZOBMLKI1070001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
AMC (7-Amino-4-methylcoumarin) standard
Catalog # ENZOBMLKI1070001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Description:
    AMC (7-Amino-4-methylcoumarin) standard
  • Seq:
    02
  • Formulation:
    30 µM in 50 mM HEPES, pH 7,4, 100 mM sodium chloride, 0,1% CHAPS, 10 mM dithiothreitol, 1 mM EDTA, 10% glycerol.

Specifications

About this item

Fluorogenic chromophore. Ex(max): 380nm, Em(max): 460nm. Reference compound.