ContactUsLinkComponent
EpiXtract® Nuclear protein isolation kit
EpiXtract® Nuclear protein isolation kit
Catalog # ENZOENZ450160100
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
undefined
EpiXtract® Nuclear protein isolation kit
Catalog # ENZOENZ450160100
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    EpiXtract® nuclear protein isolation kit
  • Seq:
    02
  • Environmentally Preferable:
Specifications
About this item
Rapid, reliable method to isolate nuclear proteins in mammalian cells and tissues.
For the extraction of nuclear proteins from mammalian cells or tissues.
EpiXtract® Nuclear Protein Isolation Kit II provides a simple and reliable method to isolate nuclear proteins. Extracts can be used for a variety of downstream applications including western blotting, protein-DNA binding assays, nuclear enzyme assays and others requiring nuclear proteins.
Ordering information: Contains Pre-extraction Buffer, Extraction Buffer, Protease Inhibitor Cocktail, DTT.