ContactUsLinkComponent
EpiXtract® total histone extraction kit (high throughput)
EpiXtract® total histone extraction kit (high throughput)
Catalog # ENZOENZ450130002
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
EpiXtract® total histone extraction kit (high throughput)
Catalog # ENZOENZ450130002
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    EpiXtract® total histone extraction kit (high throughput)
  • Seq:
    02
Specifications
About this item
Rapid, reliable, high throughput method to isolate histone proteins in mammalian cells for histone modification applications.
For the extraction of total histone proteins from mammalian cells in a 96 well plate format.
EpiXtract® Total Histone Extraction Kit (High Throughput) provides a simple and reliable method to isolate histone proteins in one hour. Extracts can be used for a variety of downstream applications such as post-translational modification assays.
Ordering information: Contains Pre-Lysis Buffer, Lysis Buffer, Balance Buffer, DTT .