ContactUsLinkComponent
Cathepsin S Assay Buffer
Cathepsin S Assay Buffer
Catalog # ENZOBMLKI4310020
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Cathepsin S Assay Buffer
Catalog # ENZOBMLKI4310020
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    20 mL
  • Description:
    Cathepsin S assay buffer
  • Seq:
    02
Specifications
About this item
50mM potassium phosphate buffer, pH 6,5, 50 mM NaCl, 2 mM EDTA, 0,01% Triton X-100®, 0,5 mM DTT.