Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Assay Buffers
MMP-1 colorimetric assay buffer
MMP-1 colorimetric assay buffer
MMP-1 colorimetric assay buffer
Catalog # ENZOBMLKI1730020
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
MMP-1 colorimetric assay buffer
Catalog # ENZOBMLKI1730020
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
20 mL
Description:
MMP-1 colorimetric assay buffer
Seq:
02
Specifications
About this item
Product Details
Product Details