Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Assay Buffers
HDAC assay buffer
HDAC assay buffer
HDAC assay buffer
Catalog # ENZOBMLKI1430020
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
HDAC assay buffer
Catalog # ENZOBMLKI1430020
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
20 mL
Description:
HDAC assay buffer
Seq:
02
Specifications
About this item
Product Details
Product Details