0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Buffers
Assay Buffers
PTP1B 2x assay buffer
PTP1B 2x assay buffer
ENZOBMLKI1310020
:
ENZO LIFE SCIENCES
:
PTP1B 2x assay buffer
ENZOBMLKI1310020
:
ENZO LIFE SCIENCES
:
Pk:
20 mL
Description:
PTP1B 2x assay buffer
Seq:
02