Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Fixation/Permeabilisation, (buffer)

Fixation/Permeabilization, (buffer), 100 mL
Catalog # ENZOADI950010100
Supplier:   Enzo Life Sciences
Fixation/Permeabilisation, (buffer)
Catalog # ENZOADI950010100
Supplier:  Enzo Life Sciences
CAS Number:  
Fixation/Permeabilisation, (buffer)

Fixation/Permeabilization, (buffer), 100 mL

CAS Number:  
Price
170,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Fixation/Permeabilization, (buffer)
  • Cat. No.:
    ENZOADI950010100
  • Formulation:
    Liquid. In PBS, pH 7,2, containing 4% formaldehyde and 0,1% Saponin.

Specifications