Order Entry
Austria
Orders LinkContactUsLinkComponent
Wash buffer concentrate

Wash buffer concentrate

Wash buffer concentrate, 100 mL
Catalog # ENZOADI801287
Supplier:   Enzo Life Sciences
Wash buffer concentrate
Catalog # ENZOADI801287
Supplier:  Enzo Life Sciences
CAS Number:  
Wash buffer concentrate

Wash buffer concentrate, 100 mL

CAS Number:  
Price
163,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Liquid. TRIS buffered saline containing detergents.

For ELISA.

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Wash buffer concentrate
  • Cat. No.:
    ENZOADI801287
  • Formulation:
    Liquid, TRIS buffered saline containing detergents

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.