Order Entry
Austria
ContactUsLinkComponent
Cyanine 5-UTP (enhanced)
Cyanine 5-UTP (enhanced)
Catalog # ENZOENZ42506
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Cyanine 5-UTP (enhanced)
Catalog # ENZOENZ42506
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    250 Nmo
  • Description:
    Cyanine 5-UTP (Enhanced) 250 nmol
  • Seq:
    02
  • Purity detail:
    Purified by ion-exchange chromatography
  • Purity:
    ≥93% (HPLC)
  • Concentration:
    10 mM
  • Formulation:
    Liquid. Solution in water.

Specifications

About this item

Cyanine 5-UTP (Enhanced) can replace UTP as a substrate for T7 RNA polymerase in labeling systems that generate labeled probes through in vitro transcription. Probes generated by these methods are suitable for multicolor fluorescence analysis, specifically dual-color expression arrays in conjunction with cyanine 3-UTP.