BK virus BioProbe®
ENZOENZ40848
New Product
:
- Pk:2 µG
- Application:
- Description:BK virus BioProbe®
- Seq:02
- Environmentally Preferable:
- Concentration:20 µg/ml
- Formulation:Liquid. In 10 mM TRIS HCl, pH 7,5, containing 1 mM EDTA.
- Size:2 µg
The BK Virus BioProbe® labeled probe is prepared by nick translation of a clone of the entire BK virus genome. BK virus, a simian polyoma virus, is related to SV40 and also bears some similarity to the human virus JC virus. BK virus is highly tumorigenic in hamsters. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).