Order Entry
Austria
ContactUsLinkComponent
Cytomegalovirus BioProbe®
Catalog # ENZOENZ40835
Supplier:  Enzo Life Sciences
CAS Number:  
Cytomegalovirus BioProbe®
Catalog # ENZOENZ40835
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    2 µG
  • Description:
    Cytomegalovirus BioProbe®
  • Seq:
    02
  • Concentration:
    20 µg/ml
  • Formulation:
    Liquid. In 10 mM TRIS HCl, pH 7,5, containing 1 mM EDTA.
  • Size:
    2 µg

Specifications

About this item

The Cytomegalovirus (CMV) BioProbe® labeled probe is prepared by nick translation of cloned fragments of the CMV Towne strain. The fragments include a total of 30-31 kb of DNA, approximately 20% of the CMV genome. The probe is specific for cytomegalovirus DNA. It does not hybridize to DNAs of other herpesviruses (i.e., herpes simplex virus or Epstein-Barr virus) but it has been found to hybridize to the DNA of CMV clinical isolates. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).