Order Entry
Austria
ContactUsLinkComponent
 
Proteasome 20S Assay Kit
Catalog # ENZOBMLAK7400001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Proteasome 20S Assay Kit
Catalog # ENZOBMLAK7400001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Proteasome 20S assay kit
  • Seq:
    02
  • Storage conditions:
    –20 °C (long term)
  • Application:
    Fluorescent detection, HTS activity assay
  • Size:
    96 well

Specifications

About this item

A QuantiZyme™ Assay System. Fluorogenic, non-radioactive assay designed to measure chymotrypsin-like protease activity of purified 20S proteasome. This 96-well assay is useful for screening inhibitors and modulators of the 20S proteasome.

The Suc-LLVY-AMC fluorogenic peptide substrate supplied with this kit is a proteasome substrate commonly used for measuring chymotrypsin-like peptidase activity. The detection of proteolytic activity is based on the release of free AMC fluorophore and offers the following advantages: non-radioactive, convenient real-time kinetics of substrate cleavage, and excellent sensitivity. A proteasome inhibitor, epoxomicin, is included as a test inhibitor for screening purposes.

Delivery: Kit includes Proteasome 20S (human) (purified), Suc-Leu-Leu-Val-Tyr-AMC, Proteasome Assay Buffer, Sodium Dodecyl Sulfate, Epoxomicin, AMC (7-Amino-4-methylcoumarin) Calibration Standard, Half Volume White Microplates.