Caspase-3 fluorometric assay kit
Catalog # ENZOALX850216KI01
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:25 Tests
- Description:Caspase-3 fluorometric assay kit
- Seq:02
- Shipping conditions:−20 °C
- Application:Fluorescent Detection, Activity Assay
- Size:25 tests
Specifications
About this item
Based on the detection of AFC (7-amino-4-trifluoromethyl coumarin) after cleavage from the labelled substrate DEVD-AFC.
Delivery: Cell Lysis Buffer (25 ml), 2X Reaction Buffer (2 ml), 1 mM DEVD-AFC (125 µl), 1 M DTT (100 µl).