Order Entry
Austria
ContactUsLinkComponent
Caspase-2 fluorometric assay kit
Caspase-2 fluorometric assay kit
Catalog # ENZOALX850214KI01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Caspase-2 fluorometric assay kit
Catalog # ENZOALX850214KI01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 Tests
  • Description:
    Caspase-2 fluorometric assay kit
  • Seq:
    02
  • Shipping conditions:
    −20 °C
  • Application:
    Fluorescent Detection, Activity Assay
  • Size:
    25 tests

Specifications

About this item

Based on the detection of AFC (7-amino-4-trifluoromethyl coumarin) after cleavage from the labelled substrate VDVAD-AFC.

Delivery: Kit includes Cell Lysis buffer (25 ml), 2X Reaction Buffer (2 ml), 1 mM VDVAD-AFC Substrate (125 µl), DTT (100 µl).