Order Entry
Austria
ContactUsLinkComponent
ProteoStat™ PDI assay kit
ProteoStat™ PDI assay kit
Supplier:  ENZO LIFE SCIENCES
Certificates
ProteoStat™ PDI assay kit
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Storage conditions:
    –80 °C (Long Term)
  • Application:
    Fluorescent Detection

Specifications

Frequently Bought Together