Order Entry
Austria
ContactUsLinkComponent
Matrix Metalloproteinase-20 (MMP-20) colorimetric drug discovery kit
Matrix Metalloproteinase-20 (MMP-20) colorimetric drug discovery kit
Catalog # ENZOBMLAK4220001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Matrix Metalloproteinase-20 (MMP-20) colorimetric drug discovery kit
Catalog # ENZOBMLAK4220001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Matrix Metalloproteinase-20 (MMP-20) O79:O98 drug discovery kit
  • Seq:
    02
  • Storage conditions:
    −80 °C
  • Application:
    Colorimetric detection, HTS activity assay
  • Size:
    1 kit

Specifications

About this item

A QuantiZyme™ Assay System. The MMP-20 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-20 (MMP-20, enamelysin) using a thiopeptide as a chromogenic substrate.

Delivery: Kit contains MMP-20 enzyme (human) (recombinant), substrate, a prototypic control inhibitor (NNGH3), assay buffer, 96-well microplate and a detailed instruction booklet.