Matrix Metalloproteinase-14 (MMP-14) colorimetric drug discovery kit
Catalog # ENZOBMLAK4160001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Description:Matrix Metalloproteinase-14 (MMP-14) colorimetric drug discovery kit
- Seq:02
- Storage conditions:−80 °C
- Application:Colorimetric detection, HTS activity assay
- Size:1 kit
Specifications
About this item
A QuantiZyme™ Assay System. The MMP-14 Colorimetric Drug Discovery Kit is a complete assay sytem designed to screen for inhibitors of matrix metalloproteinase-14 (MMP-14, membrane-type matrix metalloproteinase 1, MT1-MMP) using a thiopeptide as a chromogenic substrate.
Delivery: Kit contains MMP-14 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.