Order Entry
Austria
ContactUsLinkComponent
Matrix Metalloproteinase-13 (MMP-13) colorimetric drug discovery kit
Artikelnr. ENZOBMLAK4120001
Lieferant:  ENZO LIFE SCIENCES
CAS-Nummer:  
Matrix Metalloproteinase-13 (MMP-13) colorimetric drug discovery kit
Artikelnr. ENZOBMLAK4120001
Lieferant:  ENZO LIFE SCIENCES
CAS-Nummer:  

Spezifikationen

  • Pk:
    1 KIT
  • Description:
    Matrix Metalloproteinase-13 (MMP-13) colorimetric drug discovery kit
  • Seq:
    02
  • Storage conditions:
    −80 °C
  • Application:
    Colorimetric detection, HTS activity assay
  • Size:
    1 kit

Spezifikationen

Über diesen Artikel

A QuantiZyme™ Assay System. The MMP-13 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-13 (MMP-13, collagenase-3) using a thiopeptide as a chromogenic substrate.

Delivery: Kit contains MMP-13 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.