Matrix Metalloproteinase-9 (MMP-9) fluorometric drug discovery kit
ENZOBMLAK4110001
:
- Pk:1 KIT
- Description:Matrix Metalloproteinase-9 (MMP-9) fluorometric drug discovery kit
- Seq:02
- Storage conditions:−80 °C
- Application:Fluorescent detection, HTS activity assay
- Size:1 kit
A QuantiZyme™ Assay SystemThe MMP-9 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-9 (MMP-9, gelatinase B) using a quenched fluorogenic peptide: OmniMMP™ fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties.
: Kit contains MMP-9 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.