Matrix Metalloproteinase-7 (MMP-7) colorimetric drug discovery kit
ENZOBMLAK4060001
:
- Pk:1 KIT
- Description:Matrix Metalloproteinase-7 (MMP-7) colorimetric drug discovery kit
- Seq:02
- Storage conditions:−80 °C
- Application:Colorimetric detection, HTS activity assay
- Size:1 kit
A QuantiZyme™ Assay SystemThe MMP-7 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-7 (MMP-7, matrilysin) using a thiopeptide as a chromogenic substrate.
: Kit contains MMP-7 Enzyme (human) (recombinant), Substrate, Prototypic Control Inhibitor (NNGH), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.