Matrix Metalloproteinase-3 (MMP-3) fluorometric drug discovery kit
Catalog # ENZOBMLAK4010001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Description:Matrix Metalloproteinase-3 (MMP-3) fluorometric drug discovery kit
- Seq:02
- Storage conditions:−80 °C
- Application:Colorimetric detection, HTS activity assay
- Size:1 kit
Specifications
About this item
A QuantiZyme™ Assay System. The MMP-3 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-3 (MMP-3, stromelysin-1) using a quenched fluorogenic peptide: OmniMMP™ fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties.
Delivery: Kit contains MMP-3 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.