Order Entry
Austria
ContactUsLinkComponent
CV-Cathepsin K detection kit
CV-Cathepsin K detection kit
Catalog # ENZOBMLAK1260001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
CV-Cathepsin K detection kit
Catalog # ENZOBMLAK1260001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    CV-Cathepsin K detection kit
  • Seq:
    02
  • Storage conditions:
    4 °C
  • Application:
    Fluorescence microscopy, fluorescent detection
  • Size:
    1 kit

Specifications

About this item

Based on the dye cresyl violet and the cathepsin K LeuArg peptide recognition sequence, this kit allows the detection of cathepsin K activity in living cells by fluorescence microscopy.

Once cells are grown, staining is completed in just a few minutes. Hoechst 33342 stain and Acridine Orange are also included to allow visualization of nuclei and lysosomes.

Elevated cathepsin enzyme activity often indicates the presence of a pathological condition including cancer, Alzheimer's disease, autoimmune disease, and osteoporosis.

Ordering information: Kit contains CV-(LR)2 reagent, Acridine Orange, Hoechst 33342 stain.