Order Entry
Austria
ContactUsLinkComponent
CV-Cathepsin B detection kit
CV-Cathepsin B detection kit
  ENZOBMLAK1250001
 :  
CV-Cathepsin B detection kit
  ENZOBMLAK1250001
 :  

 

  • Pk:
    1 KIT
  • Description:
    CV-Cathepsin B detection kit
  • Seq:
    02
  • Storage conditions:
    4 °C
  • Application:
    Fluorescence microscopy, fluorescent detection
  • Size:
    1 kit

 

 

Based on the dye cresyl violet and the cathepsin B ArgArg peptide recognition sequence, this kit allows the detection of cathepsin B activity in living cells by fluorescence microscopy.

Once cells are grown, staining is completed in just a few minutes. Hoechst 33342 stain and Acridine Orange are also included to allow visualization of nuclei and lysosomes.

Elevated cathepsin enzyme activity often indicates the presence of a pathological condition including cancer, Alzheimer's disease, autoimmune disease, and osteoporosis.

 : Kit contains CV-(RR)2 reagent, Acridine Orange, Hoechst 33342 stain